missing translation for 'onlineSavingsMsg'
Learn More
3,5-Di-tert-butyl-4-hydroxybenzaldehyde, 98+%, Thermo Scientific Chemicals Product Code.: 11402568

3,5-Di-tert-butyl-4-hydroxybenzaldehyde, 98+%, Thermo Scientific Chemicals

Product Code. 11402568
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11402568

Brand: Thermo Scientific Alfa Aesar A15864.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

3,5-Di-tert-butyl-4-hydroxybenzaldehyde is employed as an OLED materials. It is also used as a pharmaceutical intermediate. The derivatives of this compound possess equipotent anti-inflammatory activities to indomethacin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Di-tert-butyl-4-hydroxybenzaldehyde is employed as an OLED materials. It is also used as a pharmaceutical intermediate. The derivatives of this compound possess equipotent anti-inflammatory activities to indomethacin.

Solubility
Soluble in hot methanol.

Notes
Air sensitive. Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1620-98-0
Molecular Formula C15H22O2
Molecular Weight (g/mol) 234.339
MDL Number MFCD00008826
InChI Key DOZRDZLFLOODMB-UHFFFAOYSA-N
Synonym 3,5-di-tert-butyl-4-hydroxybenzaldehyde, 3,5-di-t-butyl-4-hydroxybenzaldehyde, unii-95vti93vul, benzaldehyde, 3,5-bis 1,1-dimethylethyl-4-hydroxy, benzaldehyde, 3,5-di-tert-butyl-4-hydroxy, 95vti93vul, 3,5-ditert-butyl-4-hydroxy-benzaldehyde, 3,5-di tert-butyl-4-hydroxybenzaldehyde, 3,5-di-tert-butyl-4-hydroxy-benzaldehyde, 3,5-bis tert-butyl-4-hydroxybenzaldehydeShow MoreShow Less
PubChem CID 73219
IUPAC Name 3,5-ditert-butyl-4-hydroxybenzaldehyde
SMILES CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)C=O

Specifications

Melting Point 187°C to 189°C
Quantity 25 g
Beilstein 982526
Sensitivity Air Sensitive
Solubility Information Soluble in hot methanol.
Formula Weight 234.34
Percent Purity ≥98%
Chemical Name or Material 3,5-Di-tert-butyl-4-hydroxybenzaldehyde

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10245807 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog NumberA15864.14,A15864.06,A15864.22
Lot Number10241342 Certificate TypeCertificate of Analysis Date22/06/2022 Catalog NumberA15864.14,A15864.06,A15864.22
Lot Number10185497 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15864.14,A15864.06,A15864.22
Lot Number10137534 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15864.14,A15864.06,A15864.22
Lot Number10106807 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15864.14,A15864.06,A15864.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,5-Di-tert-butyl-4-hydroxybenzaldehyde, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.