missing translation for 'onlineSavingsMsg'
Learn More
3,5-Diaminobenzoic acid, 98%, Thermo Scientific Chemicals Product Code.: 10528290

3,5-Diaminobenzoic acid, 98%, Thermo Scientific Chemicals

Product Code. 10528290
500 g, Plastic bottle
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10528290

Brand: Thermo Scientific Acros 112115000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 535-87-5
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.15
MDL Number MFCD00007807
InChI Key UENRXLSRMCSUSN-UHFFFAOYSA-N
Synonym benzoic acid, 3,5-diamino, 1-carboxy-3,5-diaminobenzene, ccris 2885, 3,5-diamino benzoic acid, 3,5-diaminobenzoicacid, dabz, pubchem21025, 3,5-diaminebenzoicacid, acmc-1awxi, 3,5-diamino-benzoic acid
PubChem CID 12062
IUPAC Name 3,5-diaminobenzoic acid
SMILES C1=C(C=C(C=C1N)N)C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 235°C
Color White to Gray
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Plastic bottle
Linear Formula (H2N)2C6H3CO2H
Quantity 500 g
Beilstein 14, 453
Merck Index 15, 2980
Solubility Information Solubility in water: 7g/L (22°C). Other solubilities: soluble in alcohol and ether
Formula Weight 152.15
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3, 5-Diaminobenzoic acid, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464045 Certificate TypeCertificate of Analysis Date04/07/2024 Catalog Number112111000,112115000
Lot NumberA0401736 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number112111000,112115000
Lot NumberA0412979 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number112111000,112115000
Lot NumberA0408162 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number112111000,112115000
Lot NumberA0405863 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number112111000,112115000
compliance-icons
Product Identifier
  • 3,5-Diaminobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,5-Diaminobenzoic acid, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.