Learn More
3-(Aminomethyl)benzeneboronic acid hydrochloride, 96%, Thermo Scientific Chemicals
Description
3-(Aminomethyl)benzeneboronic acid hydrochloride is used as intermediates. Hydroxybenzene boronic acids are involved in suzuki miyaura reactions. It is a arylboronic acid. 3-Fluoro-4-hydroxybenzeneboronic acid, can be an effective catalyst for amidation and esterification of carboxylic acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3-(Aminomethyl)benzeneboronic acid hydrochloride is used as intermediates. Hydroxybenzene boronic acids are involved in suzuki miyaura reactions. It is a arylboronic acid. 3-Fluoro-4-hydroxybenzeneboronic acid, can be an effective catalyst for amidation and esterification of carboxylic acids.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Refrigerate.

Specifications
Specifications
Melting Point | 248°C to 250°C |
Quantity | 1 g |
Solubility Information | Slightly soluble in water. |
Formula Weight | 187.43 |
Percent Purity | 96% |
Chemical Name or Material | 3-(Aminomethyl)benzeneboronic acid hydrochloride |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
W14J018 | Certificate of Analysis | 04/10/2023 | H53284.03,H53284.MD |
M19I023 | Certificate of Analysis | 12/04/2022 | H53284.03,H53284.MD |
10169733 | Certificate of Analysis | 07/12/2021 | H53284.03,H53284.MD |
X15H038 | Certificate of Analysis | 07/12/2021 | H53284.03,H53284.MD |
X22G023 | Certificate of Analysis | 07/12/2021 | H53284.03,H53284.MD |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.