Learn More
Thermo Scientific Chemicals 3-Bromo-5-fluorobenzaldehyde, 95%
Form: solid
89.04 EUR - 1045.00 EUR
Chemical Identifiers
CAS | 188813-02-7 |
---|---|
Molecular Formula | C7H4BrFO |
Molecular Weight (g/mol) | 203.01 |
MDL Number | MFCD04116319 |
InChI Key | MEFQRXHVMJPOKZ-UHFFFAOYSA-N |
Synonym | 3-fluoro-5-bromobenzaldehyde, benzaldehyde, 3-bromo-5-fluoro, 5-bromo-3-fluorobenzaldehyde, 3-bromo-5-fluoro-benzaldehyde, pubchem1420, acmc-209ers, ksc174q7p, 3-bromo-5-fluorobenzaldehyde |
PubChem CID | 21986246 |
IUPAC Name | 3-bromo-5-fluorobenzaldehyde |
SMILES | C1=C(C=C(C=C1F)Br)C=O |
Description
3-Bromo-5-fluorobenzaldehyde is an intermediate in organic syntheses. Reatant in the synthesis of 3,4-diamino-3-cyclobutene-1,2-diones, 9,10-Dihydro-9,10-diboraanthracenes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Bromo-5-fluorobenzaldehyde is an intermediate in organic syntheses. Reatant in the synthesis of 3,4-diamino-3-cyclobutene-1,2-diones, 9,10-Dihydro-9,10-diboraanthracenes.
Solubility
Slightly soluble in water.
Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light, air , heat.
Chemical Identifiers
188813-02-7 | |
203.01 | |
MEFQRXHVMJPOKZ-UHFFFAOYSA-N | |
21986246 | |
C1=C(C=C(C=C1F)Br)C=O |
C7H4BrFO | |
MFCD04116319 | |
3-fluoro-5-bromobenzaldehyde, benzaldehyde, 3-bromo-5-fluoro, 5-bromo-3-fluorobenzaldehyde, 3-bromo-5-fluoro-benzaldehyde, pubchem1420, acmc-209ers, ksc174q7p, 3-bromo-5-fluorobenzaldehyde | |
3-bromo-5-fluorobenzaldehyde |
Specifications
188813-02-7 | |
1.43 g/mL | |
MFCD04116319 | |
Light sensitive | |
Slightly soluble in water. | |
C1=C(C=C(C=C1F)Br)C=O | |
203.01 | |
203.01 | |
Solid |
41°C to 43°C | |
C7H4BrFO | |
13904428 | |
3-fluoro-5-bromobenzaldehyde, benzaldehyde, 3-bromo-5-fluoro, 5-bromo-3-fluorobenzaldehyde, 3-bromo-5-fluoro-benzaldehyde, pubchem1420, acmc-209ers, ksc174q7p, 3-bromo-5-fluorobenzaldehyde | |
MEFQRXHVMJPOKZ-UHFFFAOYSA-N | |
3-bromo-5-fluorobenzaldehyde | |
21986246 | |
95% | |
3-Bromo-5-fluorobenzaldehyde |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.