missing translation for 'onlineSavingsMsg'
Learn More
3-Bromo-5-fluorobenzeneboronic acid, 95%, Thermo Scientific Chemicals Product Code.: 15429657

3-Bromo-5-fluorobenzeneboronic acid, 95%, Thermo Scientific Chemicals

Product Code. 15429657
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15429657

Brand: Thermo Scientific Alfa Aesar H52878.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

3-Bromo-5-fluorobenzeneboronic acid reactant for preparation of functionalized dihalophenylboronic acid via lithiation of dihalophenyl dioxazaborocines and reaction with electrophiles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-5-fluorobenzeneboronic acid reactant for preparation of functionalized dihalophenylboronic acid via lithiation of dihalophenyl dioxazaborocines and reaction with electrophiles.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 849062-37-9
Molecular Formula C6H5BBrFO2
Molecular Weight (g/mol) 218.82
MDL Number MFCD06411356
InChI Key UVKKALLRVWTDOY-UHFFFAOYSA-N
Synonym 3-bromo-5-fluorophenyl boronic acid, 3-bromo-5-fluorobenzeneboronic acid, 3-bromo-5-fluoro-phenyl boronic acid, acmc-209pyp, 3-bromo-5-fluorobenzeneboronic acid 95, boronic acid,b-3-bromo-5-fluorophenyl, 3-bromanyl-5-fluoranyl-phenyl boronic acid, 3-bromo-5-fluorophenylboronic acid, boronic acid, b-3-bromo-5-fluorophenyl
PubChem CID 16217688
IUPAC Name (3-bromo-5-fluorophenyl)boronic acid
SMILES OB(O)C1=CC(F)=CC(Br)=C1

Specifications

Melting Point >300°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 218.82
Percent Purity 95%
Chemical Name or Material 3-Bromo-5-fluorobenzeneboronic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Bromo-5-fluorobenzeneboronic acid, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.