missing translation for 'onlineSavingsMsg'
Learn More
3-Chloro-5-fluorobenzylamine hydrochloride, Thermo Scientific™ Product Code.: 15488007

3-Chloro-5-fluorobenzylamine hydrochloride, Thermo Scientific™

Product Code. 15488007
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15488007

Brand: Thermo Scientific Alfa Aesar H51131.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90389-34-7
Molecular Formula C7H8Cl2FN
Molecular Weight (g/mol) 196.046
MDL Number MFCD11113045
InChI Key SXKXBIIFCYGVBL-UHFFFAOYSA-N
Synonym 3-chloro-5-fluorobenzylamine hydrochloride, chembl13500, 3-chloro-5-fluorophenyl methanamine hydrochloride, 1-3-chloro-5-fluorophenyl methanamine hydrochlorideShow MoreShow Less
PubChem CID 49795983
IUPAC Name (3-chloro-5-fluorophenyl)methanamine;hydrochloride
SMILES C1=C(C=C(C=C1F)Cl)CN.Cl

Specifications

Quantity 5 g
Beilstein 5620322
Sensitivity Air Sensitive
Formula Weight 196.05
Chemical Name or Material 3-Chloro-5-fluorobenzylamine hydrochloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberG08T001 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Chloro-5-fluorobenzylamine hydrochloride, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.