missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-5-(pentafluorothio)benzylamine, 97%, Thermo Scientific™

94.52 EUR - 291.00 EUR

Chemical Identifiers

CAS 1240257-82-2
Molecular Formula C7H7ClF5NS
Molecular Weight (g/mol) 267.642
MDL Number MFCD16652457
InChI Key VUWNYLVTMIHGGJ-UHFFFAOYSA-N
Synonym 3-chloro-5-pentafluorosulfur benzylamine, 1-3-chloro-5-pentafluoro-??-sulfanyl phenyl methanamine, 3-chloro-5-pentafluorothio benzylamine
PubChem CID 66523536
IUPAC Name [3-chloro-5-(pentafluoro-$l^{6}-sulfanyl)phenyl]methanamine
SMILES C1=C(C=C(C=C1S(F)(F)(F)(F)F)Cl)CN
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15456087
SDS View Documents
Thermo Scientific Alfa Aesar
H33271.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15446087
SDS View Documents
Thermo Scientific Alfa Aesar
H33271.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Chloro-5-(pentafluorothio)benzylamine, 97%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.