missing translation for 'onlineSavingsMsg'
Learn More

3-Cyano-1-propylboronic acid pinacol ester, 97%, Thermo Scientific™

65.90 EUR - 211.00 EUR

Chemical Identifiers

CAS 238088-16-9
Molecular Formula C10H18BNO2
Molecular Weight (g/mol) 195.07
MDL Number MFCD09953480
InChI Key LVMSRWXPZLRTIC-UHFFFAOYSA-N
Synonym 3-cyano-1-propylboronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl butanenitrile, 4-tetramethyl-1,3,2-dioxaborolan-2-yl butanenitrile, 1,3,2-dioxaborolane-2-butanenitrile, 4,4,5,5-tetramethyl, 3-cyanoprop-1-ylboronic acid, pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl butyronitrile, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl butanenitrile
PubChem CID 11183176
IUPAC Name 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)butanenitrile
SMILES CC1(C)OB(CCCC#N)OC1(C)C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10165857
SDS View Documents
Thermo Scientific Acros
430530010
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10207517
SDS View Documents
Thermo Scientific
430530050
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Cyano-1-propylboronic acid pinacol ester, 97%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.