missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-5-(trifluoromethyl)benzylamine, 97%, Thermo Scientific™

45.11 EUR - 164.00 EUR

Chemical Identifiers

CAS 150517-77-4
Molecular Formula C8H7F4N
Molecular Weight (g/mol) 193.145
MDL Number MFCD00061163
InChI Key ADHUUMWSWNPEMZ-UHFFFAOYSA-N
Synonym 3-fluoro-5-trifluoromethyl benzylamine, 3-fluoro-5-trifluoromethyl phenyl methanamine, 3-fluoro-5-trifluoromethylbenzylamine, 3-fluoro-5-trifluoromethyl-benzylamine, benzenemethanamine, 3-fluoro-5-trifluoromethyl, 1-3-fluoro-5-trifluoromethyl phenyl methanamine, 5-fluoro-3-trifluoromethyl phenyl methylamine, acmc-1c7bq, ksc495o0tShow MoreShow Less
PubChem CID 2733304
IUPAC Name [3-fluoro-5-(trifluoromethyl)phenyl]methanamine
SMILES C1=C(C=C(C=C1C(F)(F)F)F)CN
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11373016
SDS View Documents
Thermo Scientific Alfa Aesar
L10836.03
1 g N/A N/A N/A
11383016
SDS View Documents
Thermo Scientific Alfa Aesar
L10836.06
5 g N/A N/A N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Fluoro-5-(trifluoromethyl)benzylamine may be employed as amine to generate different trityl-based pro-tags, by treating with activated carboxyl group-containing trityl synthons.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H227-H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Fluoro-5-(trifluoromethyl)benzylamine, 97%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.