Learn More
3-Hydroxy-4-methoxybenzoic acid, 99%, Thermo Scientific Chemicals
35.30 EUR - 99.90 EUR
Chemical Identifiers
CAS | 645-08-9 |
---|---|
Molecular Formula | C8H8O4 |
Molecular Weight (g/mol) | 168.15 |
MDL Number | MFCD00002507 |
InChI Key | LBKFGYZQBSGRHY-UHFFFAOYSA-N |
Synonym | isovanillic acid, acide isovanillique, 3-hydroxyanisic acid, 3-hydroxy-p-anisic acid, benzoic acid, 3-hydroxy-4-methoxy, p-anisic acid, 3-hydroxy, chembl88700, 3-hydroxy-4-methoxy-benzoic acid, 3-hydroxy-4-methoxybenzoicacid, 5-carboxyguaiacol |
PubChem CID | 12575 |
ChEBI | CHEBI:63798 |
IUPAC Name | 3-hydroxy-4-methoxybenzoic acid |
SMILES | COC1=C(C=C(C=C1)C(=O)O)O |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
645-08-9 | |
168.15 | |
LBKFGYZQBSGRHY-UHFFFAOYSA-N | |
12575 | |
3-hydroxy-4-methoxybenzoic acid |
C8H8O4 | |
MFCD00002507 | |
isovanillic acid, acide isovanillique, 3-hydroxyanisic acid, 3-hydroxy-p-anisic acid, benzoic acid, 3-hydroxy-4-methoxy, p-anisic acid, 3-hydroxy, chembl88700, 3-hydroxy-4-methoxy-benzoic acid, 3-hydroxy-4-methoxybenzoicacid, 5-carboxyguaiacol | |
CHEBI:63798 | |
COC1=C(C=C(C=C1)C(=O)O)O |
Specifications
645-08-9 | |
100.0 | |
Beige to Brown-Gray | |
98.5% min. (HPLC) | |
HOC6H3(OCH3)CO2H | |
10, 393 | |
LBKFGYZQBSGRHY-UHFFFAOYSA-N | |
3-hydroxy-4-methoxybenzoic acid | |
12575 | |
168.15 | |
Powder |
98.5 | |
245°C to 250°C | |
Authentic | |
C8H8O4 | |
MFCD00002507 | |
isovanillic acid, acide isovanillique, 3-hydroxyanisic acid, 3-hydroxy-p-anisic acid, benzoic acid, 3-hydroxy-4-methoxy, p-anisic acid, 3-hydroxy, chembl88700, 3-hydroxy-4-methoxy-benzoic acid, 3-hydroxy-4-methoxybenzoicacid, 5-carboxyguaiacol | |
COC1=C(C=C(C=C1)C(=O)O)O | |
168.15 | |
CHEBI:63798 | |
99% | |
3-Hydroxy-4-methoxybenzoic acid, 99% |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 211-430-5
missing translation for 'rtecsNumber' : BZ4850000
missing translation for 'tsca' : TSCA
- 3-Hydroxy-4-methoxybenzoic acid
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.