missing translation for 'onlineSavingsMsg'
Learn More
3-Methoxy-2(1H)-pyridone, 97%, Thermo Scientific Chemicals Product Code.: 11791191

3-Methoxy-2(1H)-pyridone, 97%, Thermo Scientific Chemicals

Product Code. 11791191
1 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 11791191

Brand: Thermo Scientific 163540010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20928-63-6
Molecular Formula C6H7NO2
Molecular Weight (g/mol) 125.13
MDL Number MFCD00006015
InChI Key LKIMDXQLHFCXQF-UHFFFAOYSA-N
Synonym 3-methoxypyridin-2-ol, 2-hydroxy-3-methoxypyridine, 3-methoxy-2 1h-pyridone, 2 1h-pyridinone, 3-methoxy, 3-methoxypyridin-2 1h-one, 3-methoxypyridin-2-one, 3-methoxy-2-pyridone, 2-pyridinol, 3-methoxy, 3-methoxy-2-hydroxypyridine, methyloxypyridon
PubChem CID 88731
IUPAC Name 3-methoxy-1H-pyridin-2-one
SMILES COC1=CC=CNC1=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 113.0°C to 115.0°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 125.13
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 3-Methoxy-2(1H)-pyridone
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number341271 Certificate TypeCertificate of Analysis Date25/06/2021 Catalog Number163540050,163540010
compliance-icons
Product Identifier
  • 3-Methoxy-2(1H)-pyridone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Methoxy-2(1H)-pyridone, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.