missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-5-(trifluoromethyl)aniline, 97%, Thermo Scientific™

44.35 EUR - 142.00 EUR

Chemical Identifiers

CAS 349-55-3
Molecular Formula C8H8F3NO
Molecular Weight (g/mol) 191.153
MDL Number MFCD00000393
InChI Key VTFGJEYZCUWSAM-UHFFFAOYSA-N
Synonym 3-methoxy-5-trifluoromethyl aniline, 3-amino-5-methoxybenzotrifluoride, 5-trifluoromethyl-m-anisidine, benzenamine, 3-methoxy-5-trifluoromethyl, 3-trifluoromethyl-5-methoxybenzenamine, 3-methoxy-5-trifluoromethyl-phenylamine, 3-methoxy-5-trifluoromethyl benzenamine, 5-methoxy-alpha,alpha,alpha-trifluoro-m-toluidine, 5-methoxy-3-trifluoromethyl phenylamineShow MoreShow Less
PubChem CID 605671
IUPAC Name 3-methoxy-5-(trifluoromethyl)aniline
SMILES COC1=CC(=CC(=C1)N)C(F)(F)F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15494237
SDS View Documents
Thermo Scientific Alfa Aesar
H31637.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15484237
SDS View Documents
Thermo Scientific Alfa Aesar
H31637.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c

H302+H332-H311-H315-H319

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 206-487-8

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Methoxy-5-(trifluoromethyl)aniline, 97%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.