missing translation for 'onlineSavingsMsg'
Learn More
3-Methyl-1,2-butadiene, 97% Product Code.: 15448919

3-Methyl-1,2-butadiene, 97%

Product Code. 15448919
1 g
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy

Product Code. 15448919

Brand: Thermo Scientific Alfa Aesar L10159.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 598-25-4 | C5H8 | 68.12 g/mol

3-Methyl-1,2-butadiene has been used to study the photo-induced reaction of 1,1-dimethylallene with the cyanoarenes 1,2,4,5-tetracyanobenzene, 1,4-dicyanobenzene and 1,4-dicyanonaphthalene in the presence of methanol as nucleophile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-1,2-butadiene has been used to study the photo-induced reaction of 1,1-dimethylallene with the cyanoarenes 1,2,4,5-tetracyanobenzene, 1,4-dicyanobenzene and 1,4-dicyanonaphthalene in the presence of methanol as nucleophile.
TRUSTED_SUSTAINABILITY

Specifications

CAS 598-25-4
Melting Point -114°C
Density 0.694
Boiling Point 40°C to 41°C
Flash Point −12°C (10°F)
Molecular Formula C5H8
Linear Formula (CH3)2C=C=CH2
Refractive Index 1.419
MDL Number MFCD00008896
Quantity 1 g
UN Number UN3295
Beilstein 1697090
Sensitivity Air sensitive
Synonym 3-methyl-1,2-butadiene, 1,1-dimethylallene, 1,2-butadiene, 3-methyl, 3,3-dimethylallene, 1,1-dimethylallylene, 2-methyl-2,3-butadiene, acmc-1aqbc, ch2=c=c ch3 2, 3-methyl-1,2-butadiene 1g
Solubility Information Soluble in ether,benzene,acetone,ethanol.
InChI Key PAKGDPSCXSUALC-UHFFFAOYSA-N
SMILES CC(C)=C=C
IUPAC Name 3-methylbuta-1,2-diene
Molecular Weight (g/mol) 68.12
PubChem CID 11714
Formula Weight 68.12
Percent Purity ≥97%
Chemical Name or Material 3-Methyl-1,2-butadiene
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10123372 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL10159.03
Lot NumberF01Y021 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL10159.03
Lot NumberP06G020 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL10159.03
Lot NumberY04F012 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL10159.03
Lot NumberN02C063 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL10159.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Methyl-1,2-butadiene, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.