missing translation for 'onlineSavingsMsg'
Learn More
3-Methylbenzo[b]thiophene-2-carboxylic acid, 97%, Thermo Scientific Chemicals Product Code.: 11460488

3-Methylbenzo[b]thiophene-2-carboxylic acid, 97%, Thermo Scientific Chemicals

Product Code. 11460488
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11460488

Brand: Thermo Scientific Alfa Aesar B24624.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3133-78-6
Molecular Formula C10H8O2S
Molecular Weight (g/mol) 192.232
MDL Number MFCD01830305
InChI Key YVKLUKXESFJRCE-UHFFFAOYSA-N
Synonym 3-methylbenzo b thiophene-2-carboxylic acid, 3-methylbenzothiophene-2-carboxylic acid, acmc-20a6oy, methylbenzothiophene-carboxylic, 3-methylthianaphthene-2-carboxylic acid, 3-methyl 2-benzo b thiophenecarboxylic acid, benzo b thiophene-2-carboxylicacid, 3-methyl
PubChem CID 821868
IUPAC Name 3-methyl-1-benzothiophene-2-carboxylic acid
SMILES CC1=C(SC2=CC=CC=C12)C(=O)O

Specifications

Melting Point 241°C to 244°C
Quantity 5 g
Formula Weight 192.24
Percent Purity 97%
Chemical Name or Material 3-Methylbenzo[b]thiophene-2-carboxylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Methylbenzo[b]thiophene-2-carboxylic acid, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.