Learn More
3-Nitrobenzylamine hydrochloride, 97%, Thermo Scientific Chemicals
48.94 EUR - 158.79 EUR
Chemical Identifiers
CAS | 26177-43-5 |
---|---|
Molecular Formula | C7H8N2O2·HCl |
Molecular Weight (g/mol) | 188.62 |
MDL Number | MFCD00012858 |
InChI Key | DLZXLCHQWOZGSE-UHFFFAOYSA-N |
Synonym | 3-nitrobenzylamine hydrochloride, 3-nitrophenyl methanamine hydrochloride, 3-nitrobenzylamine hcl, 3-nitrobenzylammonium hydrochloride, benzenemethanamine, 3-nitro-, monohydrochloride, 1-3-nitrophenyl methanamine hydrochloride, pubchem7485, chembl13978, m-nitrobenzylamine hydrochloride, 3-nitrobenzyl amine hydrochlorideShow More |
PubChem CID | 2724030 |
IUPAC Name | (3-nitrophenyl)methanamine;hydrochloride |
SMILES | C1=CC(=CC(=C1)[N+](=O)[O-])CN.Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10347700
|
Thermo Scientific Acros
184730010 |
1 g | Glass bottle |
48.94 EUR
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10752611
|
Thermo Scientific Acros
184730050 |
5 g | Glass bottle |
158.79 EUR
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
26177-43-5 | |
188.62 | |
DLZXLCHQWOZGSE-UHFFFAOYSA-N | |
2724030 | |
C1=CC(=CC(=C1)[N+](=O)[O-])CN.Cl |
C7H8N2O2·HCl | |
MFCD00012858 | |
3-nitrobenzylamine hydrochloride, 3-nitrophenyl methanamine hydrochloride, 3-nitrobenzylamine hcl, 3-nitrobenzylammonium hydrochloride, benzenemethanamine, 3-nitro-, monohydrochloride, 1-3-nitrophenyl methanamine hydrochloride, pubchem7485, chembl13978, m-nitrobenzylamine hydrochloride, 3-nitrobenzyl amine hydrochlorideShow More | |
(3-nitrophenyl)methanamine;hydrochloride |
Specifications
26177-43-5 | |
100.0 | |
Tan to White | |
97% | |
O2NC6H4CH2NH2·HCl | |
12, 1076 | |
DLZXLCHQWOZGSE-UHFFFAOYSA-N | |
(3-nitrophenyl)methanamine;hydrochloride | |
2724030 | |
97% | |
3-Nitrobenzylamine hydrochloride, 97% |
96.0 | |
226°C to 231°C | |
Authentic | |
C7H8N2O2·HCl | |
MFCD00012858 | |
3-nitrobenzylamine hydrochloride, 3-nitrophenyl methanamine hydrochloride, 3-nitrobenzylamine hcl, 3-nitrobenzylammonium hydrochloride, benzenemethanamine, 3-nitro-, monohydrochloride, 1-3-nitrophenyl methanamine hydrochloride, pubchem7485, chembl13978, m-nitrobenzylamine hydrochloride, 3-nitrobenzyl amine hydrochloride | |
C1=CC(=CC(=C1)[N+](=O)[O-])CN.Cl | |
188.62 | |
188.62 | |
Solid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 247-502-8
missing translation for 'tsca' : TSCA
- 3-Nitrobenzylamine hydrochloride
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.