missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylazo-2,6-diaminopyridine hydrochloride, 98+%

Product Code. 11404957
25 g, 25g
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11404957

Brand: Thermo Scientific Alfa Aesar B21886.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 136-40-3 | C11H12ClN5 | 249.702 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136-40-3
Molecular Formula C11H12ClN5
Molecular Weight (g/mol) 249.702
MDL Number MFCD00035347
InChI Key QQBPIHBUCMDKFG-UHFFFAOYSA-N
Synonym phenazopyridine hydrochloride, phenazopyridine hcl, urodine, pyridium, pyripyridium, bisteril, diridone, mallophene, phenazodine, pyrazodineShow MoreShow Less
PubChem CID 8691
ChEBI CHEBI:71419
IUPAC Name 3-phenyldiazenylpyridine-2,6-diamine;hydrochloride
SMILES C1=CC=C(C=C1)N=NC2=C(N=C(C=C2)N)N.Cl

Specifications

Melting Point ∼240°C (decomposition)
Quantity 25 g
Formula Weight 249.7
Percent Purity ≥98%
Chemical Name or Material 3-Phenylazo-2,6-diaminopyridine hydrochloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot NumberS12J030 Certificate TypeCertificate of Analysis Date24/10/2023 Product CodeB21886.06, B21886.03, B21886.14
Lot NumberS14J039 Certificate TypeCertificate of Analysis Date21/06/2023 Product CodeB21886.06, B21886, B21886.14
Lot NumberM04I043 Certificate TypeCertificate of Analysis Date12/04/2022 Product CodeB21886.06, B21886.14
Lot NumberI16U015 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeB21886.06, B21886.14
Lot NumberI29W004 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeB21886.06, B21886.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Phenylazo-2,6-diaminopyridine hydrochloride, 98+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.