missing translation for 'onlineSavingsMsg'
Learn More

3-(tert-Butyldimethylsiloxy)thiophenol, 95%, Thermo Scientific Chemicals

Product Code. p-7047016
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11354006

Brand: Thermo Scientific Alfa Aesar L17238.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 216393-56-5
Molecular Formula C12H20OSSi
Molecular Weight (g/mol) 240.44
MDL Number MFCD01318115
InChI Key KRHCTXFJCFMFOR-UHFFFAOYSA-N
Synonym 3-tert-butyldimethylsilyl oxy benzenethiol, 3-tert-butyldimethylsiloxy thiophenol, acmc-20ani3, 3-tert-butyl-dimethylsiloxy-thiophenol, 3-tert-butyldimethylsilyloxy benzenethiol, 3-t-butyl-dimethyl-silanyloxy-benzenethiol, 3-tert-butyl-dimethyl-silanyloxy-benzenethiol, 3-tert-butyl dimethyl silyl oxy benzene-1-thiol, 3-1,1-dimethylethyl dimethyl silyl oxy benzenethiol
PubChem CID 21976868
SMILES CC(C)(C)[Si](C)(C)OC1=CC(S)=CC=C1

Specifications

Refractive Index 1.521
Quantity 1 g
Sensitivity Air Sensitive
IUPAC Name 3-[(tert-butyldimethylsilyl)oxy]benzene-1-thiol
Formula Weight 240.44
Percent Purity 95%
Chemical Name or Material 3-(tert-Butyldimethylsiloxy)thiophenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.