missing translation for 'onlineSavingsMsg'
Learn More

(3R-cis)-Tetrahydro-3-trichloromethyl-1H,3H-pyrrolo[1,2-c]oxazol-1-one, 98%, Thermo Scientific™

Product Code. 11476300
1 g, 1g
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11476300

Brand: Thermo Scientific Alfa Aesar H27164.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97538-67-5
Molecular Formula C7H8Cl3NO2
Molecular Weight (g/mol) 244.496
MDL Number MFCD08274464
InChI Key GWQBXRYSVSZLSL-UJURSFKZSA-N
Synonym 3r,7as-3-trichloromethyl tetrahydropyrrolo 1,2-c oxazol-1 3h-one, 2r,5s-2-trichloromethyl-3-oxa-1-azabicyclo 3.3.0 octan-4-one, 3r,7as-3-trichloromethyl-hexahydropyrrolo 1,2-c 1,3 oxazol-1-one, 2r,5s-2-trichloromethyl-3-oxa-1-azabicyclo 3,3,0 octane-4-one, 3r,7as-3-trichloromethyl-tetrahydro-3h-pyrrolo 1,2-c 1,3 oxazol-1-one, l-proline-chloral condensation product, 3r-trichloromethyl-cis-tetrahydropyrrolo 1,2-c oxazol-1-one, 2r,5s-2-trichloromethyl-1-aza-3-oxabicyclo 3.3.0 octan-4-one, 3r,7as-3-trichloromethyl-tetrahydro-pyrrolo 1,2-c oxazol-1-one, 3r,7as-3-trichloromethyl-tetrahydropyrrolo 1,2-c oxazol-1 3h-one
PubChem CID 10944680
IUPAC Name (3R,7aS)-3-(trichloromethyl)-5,6,7,7a-tetrahydro-3H-pyrrolo[1,2-c][1,3]oxazol-1-one
SMILES C1CC2C(=O)OC(N2C1)C(Cl)(Cl)Cl

Specifications

Melting Point 109°C (decomposition)
Quantity 1 g
Optical Rotation −2.9° (c= 1 in Chloroform)
Formula Weight 244.51
Percent Purity 98%
Chemical Name or Material (3R-cis)-Tetrahydro-3-trichloromethyl-1H,3H-pyrrolo[1,2-c]oxazol-1-one

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Product Code
Lot Number10138258 Certificate TypeCertificate of Analysis Date06/12/2021 Product Code
Lot Number10164150 Certificate TypeCertificate of Analysis Date06/12/2021 Product Code
Lot Number10157430 Certificate TypeCertificate of Analysis Date06/12/2021 Product Code
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(3R-cis)-Tetrahydro-3-trichloromethyl-1H,3H-pyrrolo[1,2-c]oxazol-1-one, 98%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.