missing translation for 'onlineSavingsMsg'
Learn More
4,4'-Azobis(4-cyanovaleric acid), 98%, cont. ca 18% water, Thermo Scientific Chemicals Product Code.: 15445136

4,4'-Azobis(4-cyanovaleric acid), 98%, cont. ca 18% water, Thermo Scientific Chemicals

Product Code. 15445136
100 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 15445136

Brand: Thermo Scientific Alfa Aesar H59611.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

4,4'-Azobis(4-cyanovaleric acid) is used as a free radical imitator in polymer synthesis such as polyvinyl chloride, polyacrylonitrile, polyvinyl alcohol and synthetic fibers. It is utilized for both heterogeneous and homogenous free-radical polymerizations, and as an initiator in reversible addition-fragmentation chain transfer polymerization (RAFT). It is also used as a blowing agent for plastics and elastomers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Azobis(4-cyanovaleric acid) is used as a free radical imitator in polymer synthesis such as polyvinyl chloride, polyacrylonitrile, polyvinyl alcohol and synthetic fibers. It is utilized for both heterogeneous and homogenous free-radical polymerizations, and as an initiator in reversible addition-fragmentation chain transfer polymerization (RAFT). It is also used as a blowing agent for plastics and elastomers.

Solubility
Soluble in water.

Notes
Light senstive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2638-94-0
Molecular Formula C12H16N4O4
Molecular Weight (g/mol) 280.284
MDL Number MFCD00002799
InChI Key VFXXTYGQYWRHJP-UHFFFAOYSA-N
Synonym 4,4'-azobis 4-cyanovaleric acid, azobis cyanovaleric acid, 4,4'-diazene-1,2-diyl bis 4-cyanopentanoic acid, 4,4'-azobis 4-cyanopentanoic acid, acva, 4,4-azobis 4-cyanovaleric acid, valeric acid, 4,4'-azobis 4-cyano, pentanoic acid, 4,4'-azobis 4-cyano, 4,4'-azobis 4-cyano valeric acid acvaShow MoreShow Less
PubChem CID 92938
IUPAC Name 4-[(4-carboxy-2-cyanobutan-2-yl)diazenyl]-4-cyanopentanoic acid
SMILES CC(CCC(=O)O)(C#N)N=NC(C)(CCC(=O)O)C#N

Specifications

Melting Point 118°C to 125°C (decomposition)
Linear Formula HOCOCH2CH2C(CH3)(CN)N=NC(CH3)(CN)CH2CH2COOH
UN Number UN1325
Beilstein 1729856
Solubility Information Soluble in water.
Quantity 100 g
Formula Weight 280.28
Sensitivity Light sensitive
Percent Purity 98%
Assay contains ≈18% water
Chemical Name or Material 4,4'-Azobis(4-cyanovaleric acid)
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN13L025 Certificate TypeCertificate of Analysis Date17/02/2025 Catalog NumberH59611.14,H59611.22
Lot NumberS08H044 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH59611.14,H59611.22
Lot NumberU28G042 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH59611.14,H59611.22
Lot NumberT07H006 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH59611.14,H59611.22
Lot NumberQ14B027 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH59611.14,H59611.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4,4'-Azobis(4-cyanovaleric acid), 98%, cont. ca 18% water, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.