Learn More
4,7-Dichloroquinoline, 98%, Thermo Scientific Chemicals Product Code.: 11408576
4,7-Dichloroquinoline, 98%, Thermo Scientific Chemicals
Product Code. 11408576
500 g
Click to view available options
Quantity:
25 g
100 g
500 g
Product Code. 11408576
Brand: Thermo Scientific Alfa Aesar A15985.36
Request Bulk or Custom Format
4,7-Dichloroquinoline is used as a drug chloroquine phosphate intermediate. It is also is used in the synthesis of hybrid aminoquinoline-triazine derivatives that show anti-microbial activity. In addition, it is used in the synthesis of novel oxazolidinones as anti-microbial agents showing efficacy against common bacterial strains.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4,7-Dichloroquinoline is used as a drug chloroquine phosphate intermediate. It is also is used in the synthesis of hybrid aminoquinoline-triazine derivatives that show anti-microbial activity. In addition, it is used in the synthesis of novel oxazolidinones as anti-microbial agents showing efficacy against common bacterial strains.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 86-98-6
Molecular Formula C9H5Cl2N
Molecular Weight (g/mol) 198.046
MDL Number MFCD00006774
InChI Key HXEWMTXDBOQQKO-UHFFFAOYSA-N
PubChem CID 6866
IUPAC Name 4,7-dichloroquinoline
SMILES C1=CC2=C(C=CN=C2C=C1Cl)Cl
Specifications
Melting Point 83°C to 87°C
Boiling Point 148°C (10 mmHg)
Flash Point 164°C (327°F)
Quantity 500 g
Beilstein 125359
Synonym quinoline, 4,7-dichloro, 4,7-dichloro-quinoline, unii-z61o2heq2j, z61o2heq2j, 4,7dichloroquinoline, pubchem5880, 4.7-dichloroquinoline, 4,7 dichloroquinoline, quinoline,7-dichloro, amodiaquine impurity 1
Solubility Information Insoluble in water.
Formula Weight 198.05
Percent Purity 98%
Chemical Name or Material 4,7-Dichloroquinoline
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10199489 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15985.14,A15985.36,A15985.22
Lot Number28520 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15985.14,A15985.36,A15985.22
Lot Number10228649 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15985.14,A15985.36,A15985.22
Lot Number10128440 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15985.14,A15985.36,A15985.22
Lot NumberGBFA008624 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15985.14,A15985.36,A15985.22
Product Title
4,7-Dichloroquinoline, 98%, Thermo Scientific Chemicals >