missing translation for 'onlineSavingsMsg'
Learn More
4-Acetoxybenzoic acid, 98+%, Thermo Scientific Chemicals Product Code.: 11329439

4-Acetoxybenzoic acid, 98+%, Thermo Scientific Chemicals

Product Code. 11329439
50 g
Click to view available options
Quantity:
25 g
50 g
250 g
500 g
This item is not returnable. View return policy

Product Code. 11329439

Brand: Thermo Scientific Alfa Aesar A11350.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2345-34-8
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.16
MDL Number MFCD00002540
InChI Key GDBUZIKSJGRBJP-UHFFFAOYSA-N
Synonym 4-acetoxybenzoic acid, p-acetoxybenzoic acid, 4-acetyloxy benzoic acid, 4-carboxyphenyl acetate, benzoic acid, 4-acetyloxy, p-hydroxybenzoic acid acetate, p-acetyloxybenzoic acid, p-carboxyphenyl acetate, benzoic acid, p-hydroxy-, acetate, 4-acetoxy benzoic acid
PubChem CID 16865
ChEBI CHEBI:86560
IUPAC Name 4-acetyloxybenzoic acid
SMILES CC(=O)OC1=CC=C(C=C1)C(O)=O

Specifications

Melting Point 192°C to 195°C
Quantity 50 g
Beilstein 1871997
Formula Weight 180.15
Percent Purity ≥98%
Chemical Name or Material 4-Acetoxybenzoic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10253304 Certificate TypeCertificate of Analysis Date04/12/2024 Catalog NumberA11350.14,A11350.18,A11350.30
Lot Number10251306 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA11350.14,A11350.18,A11350.30
Lot Number10250359 Certificate TypeCertificate of Analysis Date09/04/2024 Catalog NumberA11350.14,A11350.18,A11350.30
Lot Number10247440 Certificate TypeCertificate of Analysis Date21/07/2023 Catalog NumberA11350.14,A11350.36,A11350.18,A11350.30
Lot Number10238256 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA11350.14,A11350.18,A11350.30
compliance-icons
Product Identifier
  • 4-Acetoxybenzoic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 4-Acetoxybenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Acetoxybenzoic acid, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.