Learn More
4-Allyl-2,6-dimethoxyphenol, 98%, Thermo Scientific Chemicals Product Code.: 11419457
4-Allyl-2,6-dimethoxyphenol, 98%, Thermo Scientific Chemicals
Product Code. 11419457
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
Product Code. 11419457
Brand: Thermo Scientific Alfa Aesar B21981.03
Request Bulk or Custom Format
4-Allyl-2,6-dimethoxyphenol is used as a flavoring agent or adjuvant. Also used to produce 2,6-Dimethoxy-4-propyl-phenol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Allyl-2,6-dimethoxyphenol is used as a flavoring agent or adjuvant. Also used to produce 2,6-Dimethoxy-4-propyl-phenol.
Solubility
Soluble in fats. Insoluble in water.
Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6627-88-9
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00008655
InChI Key FWMPKHMKIJDEMJ-UHFFFAOYSA-N
Synonym 4-allyl-2,6-dimethoxyphenol, methoxyeugenol, 4-allylsyringol, phenol, 2,6-dimethoxy-4-2-propenyl, 2,6-dimethoxychavicol, phenol, 4-allyl-2,6-dimethoxy, unii-8vf00ywp89, 2,6-dimethoxy-4-allylphenol, 2,6-dimethoxy-4-prop-2-en-1-yl phenol, 4-hydroxy-3,5-dimethoxyallylbenzene
PubChem CID 226486
ChEBI CHEBI:86562
IUPAC Name 2,6-dimethoxy-4-prop-2-enylphenol
SMILES COC1=CC(CC=C)=CC(OC)=C1O
Specifications
Density 1.092
Boiling Point 168°C to 169°C (11 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.548
Quantity 1 g
Solubility Information Soluble in fats. Insoluble in water.
Formula Weight 194.23
Percent Purity 98%
Chemical Name or Material 4-Allyl-2,6-dimethoxyphenol
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10241285 Certificate TypeCertificate of Analysis Date14/06/2022 Catalog NumberB21981.03,B21981.14,B21981.06
Lot NumberI7293A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21981.03,B21981.14,B21981.06
Lot Number10157187 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21981.03,B21981.14,B21981.06
Lot Number10161278 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21981.03,B21981.14,B21981.06
Lot Number10226889 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21981.03,B21981.14,B21981.06
Product Title
4-Allyl-2,6-dimethoxyphenol, 98%, Thermo Scientific Chemicals >