missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-1,2,2,6,6-pentamethylpiperidine, 99%, Thermo Scientific Chemicals

54.67 EUR - 163.82 EUR

Chemical Identifiers

CAS 40327-96-6
Molecular Formula C10H24N2
Molecular Weight (g/mol) 172.32
MDL Number MFCD01861832
InChI Key CGXOAAMIQPDTPE-UHFFFAOYSA-P
Synonym 4-amino-1,2,2,6,6-pentamethylpiperidine, 1,2,2,6,6-pentamethyl-4-aminopiperidine, 1,2,2,6,6-pentamethyl-4-piperidinamine, 1,2,2,6,6-pentamethyl-4-piperidylamine, 4-ammo-1,2,2,6,6-pentamethylpiperidine, 4-piperidinamine,1,2,2,6,6-pentamethyl, 1,2,-2,6,6-pentamethyl-4-aminopiperidine, 1,2,2,6,6,-pentamethyl-4-aminopiperidine, 4-amino-1,2,2,6,6-pentamethyl-piperidine, 4-amino-1,2,2,6,6-pentamethyl piperidine, 97+%
PubChem CID 693792
IUPAC Name 1,2,2,6,6-pentamethylpiperidin-4-amine
SMILES C[NH+]1C(C)(C)CC([NH3+])CC1(C)C
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10546451
SDS View Documents
Thermo Scientific Acros
335940010
1 g Glass bottle
54.67 EUR
1g
Please to purchase this item. Need a web account? Register with us today!
10285911
SDS View Documents
Thermo Scientific Acros
335940050
5 g Glass bottle
163.82 EUR
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Amino-1,2,2,6,6-pentamethylpiperidine, 99%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.