missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-3-iodobenzoic acid, 98%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Resin-bound 3-[2-(acetylamino)ethyl]-2- iodo-1H-indole-5-carboxamide was generated starting from Polystyrene- Rink resin and 4-amino-3-iodobenzoic acid.
Solubility
Soluble in water(50 g/L) (25°C).
Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light. Store in the dark.
Resin-bound 3-[2-(acetylamino)ethyl]-2- iodo-1H-indole-5-carboxamide was generated starting from Polystyrene- Rink resin and 4-amino-3-iodobenzoic acid.
Solubility
Soluble in water(50 g/L) (25°C).
Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light. Store in the dark.

Specifications
Specifications
Quantity | 1 g |
Sensitivity | Light sensitive |
Solubility Information | Soluble in water(50g/L) (25°C). |
Formula Weight | 263.03 |
Percent Purity | 98% |
Chemical Name or Material | 4-Amino-3-iodobenzoic acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
4-Amino-3-iodobenzoic acid, 98%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction