missing translation for 'onlineSavingsMsg'
Learn More
4-Bromo-1-(triisopropylsilyl)-1H-indole, ≥97%, Thermo Scientific™ Product Code.: 10577863

4-Bromo-1-(triisopropylsilyl)-1H-indole, ≥97%, Thermo Scientific™

Product Code. 10577863
1 g, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10577863

Brand: Thermo Scientific Maybridge CC39510DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 412048-44-3
Molecular Formula C17H26BrNSi
Molecular Weight (g/mol) 352.391
MDL Number MFCD05664418
InChI Key ZXHMUQVMGJYGAV-UHFFFAOYSA-N
Synonym 4-bromo-1-triisopropylsilyl-1h-indole, 4-bromo-1-triisopropylsilyl indole, 4-bromo-triisopropylsilanyl-1h-indole, 4-bromo-1-triisopropylsilanyl-1h-indole, 4-bromo-1-tri-isopropylsilyl-1h-indole, 4-bromoindol-1-yl-tri propan-2-yl silane
PubChem CID 2795573
IUPAC Name (4-bromoindol-1-yl)-tri(propan-2-yl)silane
SMILES CC(C)[Si](C(C)C)(C(C)C)N1C=CC2=C1C=CC=C2Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 352.39
Percent Purity 97%
Chemical Name or Material 4-Bromo-1-(triisopropylsilyl)-1H-indole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Bromo-1-(triisopropylsilyl)-1H-indole, ≥97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.