Learn More
4-Bromobenzeneboronic acid, 97+%, Thermo Scientific Chemicals
29.75 EUR - 389.94 EUR
Chemical Identifiers
CAS | 5467-74-3 |
---|---|
Molecular Formula | C6H6BBrO2 |
Molecular Weight (g/mol) | 200.83 |
MDL Number | MFCD00002104 |
InChI Key | QBLFZIBJXUQVRF-UHFFFAOYSA-N |
Synonym | 4-bromophenyl boronic acid, 4-bromobenzeneboronic acid, 4-bromophenylboric acid, p-bromophenylboronic acid, p-bromophenylboric acid, p-bromobenzeneboronic acid, boronic acid, 4-bromophenyl, benzeneboronic acid, p-bromo, p-bromophenyl boronic acid, 4-bromo phenyl boronic acid |
PubChem CID | 79599 |
IUPAC Name | (4-bromophenyl)boronic acid |
SMILES | OB(O)C1=CC=C(Br)C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11301337
|
Thermo Scientific Alfa Aesar
L01565.03 |
1 g |
29.75 EUR
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11311337
|
Thermo Scientific Alfa Aesar
L01565.06 |
5 g |
95.48 EUR
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11321337
|
Thermo Scientific Alfa Aesar
L01565.14 |
25 g |
389.94 EUR
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4-Bromobenzeneboronic acid is a reagent used for Palladium catalyzed Suzuki-Miyaura cross-couplings, Tandem-type Pd(II)-catalyzed oxidative Heck reaction and intramolecular C-H amidation sequence. It is also used in the preparation of Protein modulators and enzymatic and kinase inhibitors, Gallate-based obovatol analogs with potential anti-tumor activity. It is used as a reagent for Copper-mediated ligandless aerobic fluoroalkylation of arylboronic acids with fluoroalkyl iodides, Pd-catalyzed arylative cyclization of alkyne-tethered enals or enones via carbopalladation of alkynes, Copper-catalyzed cross-couplings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Bromobenzeneboronic acid is a reagent used for Palladium catalyzed Suzuki-Miyaura cross-couplings, Tandem-type Pd(II)-catalyzed oxidative Heck reaction and intramolecular C-H amidation sequence. It is also used in the preparation of Protein modulators and enzymatic and kinase inhibitors, Gallate-based obovatol analogs with potential anti-tumor activity. It is used as a reagent for Copper-mediated ligandless aerobic fluoroalkylation of arylboronic acids with fluoroalkyl iodides, Pd-catalyzed arylative cyclization of alkyne-tethered enals or enones via carbopalladation of alkynes, Copper-catalyzed cross-couplings.
Solubility
Soluble in methanol. Insoluble in water.
Notes
Store in a cool and dark place. Store away from incompatible materials such as oxidizing agents. Incompatible with Oxidizing agents, Acids, Bases, Alcohols
Chemical Identifiers
5467-74-3 | |
200.83 | |
QBLFZIBJXUQVRF-UHFFFAOYSA-N | |
79599 | |
OB(O)C1=CC=C(Br)C=C1 |
C6H6BBrO2 | |
MFCD00002104 | |
4-bromophenyl boronic acid, 4-bromobenzeneboronic acid, 4-bromophenylboric acid, p-bromophenylboronic acid, p-bromophenylboric acid, p-bromobenzeneboronic acid, boronic acid, 4-bromophenyl, benzeneboronic acid, p-bromo, p-bromophenyl boronic acid, 4-bromo phenyl boronic acid | |
(4-bromophenyl)boronic acid |
Specifications
5467-74-3 | |
C6H6BBrO2 | |
2936347 | |
Soluble in methanol. Insoluble in water. | |
OB(O)C1=CC=C(Br)C=C1 | |
200.83 | |
200.83 | |
4-Bromobenzeneboronic acid |
282°C to 286°C | |
MFCD00002104 | |
4-bromophenyl boronic acid, 4-bromobenzeneboronic acid, 4-bromophenylboric acid, p-bromophenylboronic acid, p-bromophenylboric acid, p-bromobenzeneboronic acid, boronic acid, 4-bromophenyl, benzeneboronic acid, p-bromo, p-bromophenyl boronic acid, 4-bromo phenyl boronic acid | |
QBLFZIBJXUQVRF-UHFFFAOYSA-N | |
(4-bromophenyl)boronic acid | |
79599 | |
≥97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 226-779-9
missing translation for 'rtecsNumber' : CY8650000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.