Learn More
4-Bromobenzonitrile, 98+%, Thermo Scientific Chemicals
39.90 EUR - 500.49 EUR
Chemical Identifiers
CAS | 623-00-7 |
---|---|
Molecular Formula | C7H4BrN |
Molecular Weight (g/mol) | 182.02 |
MDL Number | MFCD00001811 |
InChI Key | HQSCPPCMBMFJJN-UHFFFAOYSA-N |
Synonym | p-bromobenzonitrile, benzonitrile, 4-bromo, benzonitrile, p-bromo, 1-bromo-4-cyanobenzene, 4-bromo benzonitrile, 4-cyano-bromobenzene, 4-bromo-4'-cyanobenzene, 4-cyanobromobenzene, bromobenzonitrile-4, 4-bromocyanobenzene |
PubChem CID | 12162 |
IUPAC Name | 4-bromobenzonitrile |
SMILES | BrC1=CC=C(C=C1)C#N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11484293
|
Thermo Scientific Alfa Aesar
A10914.09 |
10 g |
39.90 EUR
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11494293
|
Thermo Scientific Alfa Aesar
A10914.18 |
50 g |
167.84 EUR
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11404303
|
Thermo Scientific Alfa Aesar
A10914.30 |
250 g |
500.49 EUR
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4-Bromobenzonitrile is used as reagents and solvents. Reaction of 2-propanol, 4-bromobenzonitrile, and NaH in the presence of 1.5 mol % Pd2(dba)3 and 3 mol % (S)-(-)2,2?-bis(di-p-tolylphosphino)- 1,1?-binaphthyl at 50°C afforded 4-isopropoxy- benzonitrile in 80% isolated yield. The synthesis of 4-cyanobiphenyl at room temperature in a flow injection microreactor, using a supported catalyst, without the addition of a base gave a product yield of 67±7% (n=6). This was achieved by injecting 4-bromobenzonitrile. The 3-arylation of 2,5-dimethylthiophene with 4-bromopropiophenone, 4-trifluoromethylbromobenzene or 4-bromobenzonitrile gave 3-5 in only 38-50% yields in Palladium-catalysed direct 3-or 4-arylation of thiophene derivatives using aryl bromides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Bromobenzonitrile is used as reagents and solvents. Reaction of 2-propanol, 4-bromobenzonitrile, and NaH in the presence of 1.5 mol % Pd2(dba)3 and 3 mol % (S)-(-)2,2′-bis(di-p-tolylphosphino)- 1,1′-binaphthyl at 50°C afforded 4-isopropoxy- benzonitrile in 80% isolated yield. The synthesis of 4-cyanobiphenyl at room temperature in a flow injection microreactor, using a supported catalyst, without the addition of a base gave a product yield of 67±7% (n=6). This was achieved by injecting 4-bromobenzonitrile. The 3-arylation of 2,5-dimethylthiophene with 4-bromopropiophenone, 4-trifluoromethylbromobenzene or 4-bromobenzonitrile gave 3-5 in only 38-50% yields in Palladium-catalysed direct 3-or 4-arylation of thiophene derivatives using aryl bromides.
Solubility
Very slightly soluble in water(0.2g/L). Soluble in alcohol and ether.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
623-00-7 | |
182.02 | |
HQSCPPCMBMFJJN-UHFFFAOYSA-N | |
12162 | |
BrC1=CC=C(C=C1)C#N |
C7H4BrN | |
MFCD00001811 | |
p-bromobenzonitrile, benzonitrile, 4-bromo, benzonitrile, p-bromo, 1-bromo-4-cyanobenzene, 4-bromo benzonitrile, 4-cyano-bromobenzene, 4-bromo-4'-cyanobenzene, 4-cyanobromobenzene, bromobenzonitrile-4, 4-bromocyanobenzene | |
4-bromobenzonitrile |
Specifications
623-00-7 | |
1.562 | |
130°C (266°F) | |
MFCD00001811 | |
2041518 | |
Very slightly soluble in water(0.2g/L). Soluble in alcohol and ether. | |
BrC1=CC=C(C=C1)C#N | |
182.02 | |
182.03 | |
4-Bromobenzonitrile |
111°C to 114°C | |
235°C to 237°C | |
C7H4BrN | |
UN3439 | |
p-bromobenzonitrile, benzonitrile, 4-bromo, benzonitrile, p-bromo, 1-bromo-4-cyanobenzene, 4-bromo benzonitrile, 4-cyano-bromobenzene, 4-bromo-4'-cyanobenzene, 4-cyanobromobenzene, bromobenzonitrile-4, 4-bromocyanobenzene | |
HQSCPPCMBMFJJN-UHFFFAOYSA-N | |
4-bromobenzonitrile | |
12162 | |
≥98% |
Safety and Handling
GHS H Statement
H301-H331-H312-H315-H319-H335
Toxic if swallowed.
Toxic if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P312-P330-P363-P501c
H302+H312-H319
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
missing translation for 'einecsNumber' : 210-764-9
missing translation for 'rtecsNumber' : DI2460000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.