missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
4-Decyne is used in acid zinc plating.
Solubility
Insoluble in water.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
4-Decyne is used in acid zinc plating.
Solubility
Insoluble in water.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.

Specifications
Specifications
Density | 0.768 |
Boiling Point | 75°C to 76°C (20 mmHg) |
Flash Point | 52°C (125°F) |
Refractive Index | 1.434 |
Quantity | 5 g |
UN Number | UN3295 |
Beilstein | 1698308 |
Solubility Information | Insoluble in water. |
Formula Weight | 138.26 |
Percent Purity | 98% |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
P08521 | Certificate of Analysis | 07/12/2021 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
4-Decyne, 98%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction