missing translation for 'onlineSavingsMsg'
Learn More
4-Fluoro-2-(trifluoromethyl)benzaldehyde, 98%, Thermo Scientific™ Product Code.: 11429047

4-Fluoro-2-(trifluoromethyl)benzaldehyde, 98%, Thermo Scientific™

Product Code. 11429047
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11429047

Brand: Thermo Scientific Alfa Aesar B20621.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90176-80-0
Molecular Formula C8H4F4O
Molecular Weight (g/mol) 192.113
MDL Number MFCD00061313
InChI Key NONOHEMDNFTKCZ-UHFFFAOYSA-N
Synonym 4-fluoro-2-trifluoromethyl benzaldehyde, 2-trifluoromethyl-4-fluorobenzaldehyde, 4-fluoro-2-trifluoromethyl-benzaldehyde, benzaldehyde, 4-fluoro-2-trifluoromethyl, 5-fluoro-2-formylbenzotrifluoride, alpha,alpha,alpha,4-tetrafluoro-o-tolualdehyde, pubchem4241, ksc487q2t, paragos 530044
PubChem CID 522940
IUPAC Name 4-fluoro-2-(trifluoromethyl)benzaldehyde
SMILES C1=CC(=C(C=C1F)C(F)(F)F)C=O

Specifications

Density 1.408
Boiling Point 113°C to 115°C
Flash Point 56°C (132°F)
Refractive Index 1.453
Quantity 1 g
UN Number UN1989
Beilstein 5932091
Sensitivity Air Sensitive
Formula Weight 192.11
Percent Purity 98%
Chemical Name or Material 4-Fluoro-2-(trifluoromethyl)benzaldehyde
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Fluoro-2-(trifluoromethyl)benzaldehyde, 98%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.