missing translation for 'onlineSavingsMsg'
Learn More
4-Hexyn-3-ol, 95%, Thermo Scientific™ Product Code.: 11362237

4-Hexyn-3-ol, 95%, Thermo Scientific™

Product Code. 11362237
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11362237

Brand: Thermo Scientific Alfa Aesar L09727.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20739-59-7
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.15
MDL Number MFCD00041591
InChI Key KARLLBDFLHNKBO-UHFFFAOYNA-N
Synonym 4-hexyn-3-ol, ethyl 1-propynyl carbinol, 1-pentyn-3-ol, methyl, 4-hexyne-3-ol, acmc-1cpld
PubChem CID 140751
IUPAC Name hex-4-yn-3-ol
SMILES CCC(O)C#CC

Specifications

Density 0.894
Boiling Point 94°C to 95°C (105 mmHg)
Refractive Index 1.45
Quantity 5 g
UN Number UN1987
Beilstein 1739392
Formula Weight 98.15
Percent Purity 95%
Chemical Name or Material 4-Hexyn-3-ol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Hexyn-3-ol, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.