Learn More
4-Isopropylphenol, 98%, Thermo Scientific Chemicals Product Code.: 11324709
4-Isopropylphenol, 98%, Thermo Scientific Chemicals
Product Code. 11324709
1 kg
Click to view available options
Quantity:
50 g
250 g
1 kg
Product Code. 11324709
Brand: Thermo Scientific Alfa Aesar B22620.A1
Request Bulk or Custom Format
4-Isopropylphenol was used in the synthesis of vinyl compounds. It is used as a flavor & fragrance agent in cosmetics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Isopropylphenol was used in the synthesis of vinyl compounds. It is used as a flavor & fragrance agent in cosmetics.
Solubility
Slightly soluble in water, soluble in ethanol and ethyl ether.
Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 99-89-8
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00002372
InChI Key YQUQWHNMBPIWGK-UHFFFAOYSA-N
Synonym 4-isopropylphenol, p-isopropylphenol, p-cumenol, australol, phenol, 4-1-methylethyl, 4-1-methylethyl phenol, 1-hydroxy-4-isopropylbenzene, 4-propan-2-yl phenol, phenol, p-isopropyl, para-isopropylphenol
PubChem CID 7465
IUPAC Name 4-propan-2-ylphenol
SMILES CC(C)C1=CC=C(C=C1)O
Specifications
Melting Point 59°C to 61°C
Boiling Point 211°C to 212°C
Flash Point 108°C (226°F)
Quantity 1 kg
UN Number UN2430
Beilstein 1363564
Solubility Information Slightly soluble in water,soluble in ethanol and ethyl ether.
Formula Weight 136.2
Percent Purity 98%
Chemical Name or Material 4-Isopropylphenol
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0442281 Certificate TypeCertificate of Analysis Date22/04/2024 Catalog NumberB22620.18,B22620.30,B22620.A1
Lot NumberA0432199 Certificate TypeCertificate of Analysis Date29/12/2023 Catalog NumberB22620.18,B22620.30,B22620.A1
Lot Number10237911 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberB22620.18,B22620.30,B22620.A1
Lot Number10227275 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22620.18,B22620.30,B22620.A1
Lot Number10227061 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22620.18,B22620.30,B22620.A1
Product Title
4-Isopropylphenol, 98%, Thermo Scientific Chemicals >