missing translation for 'onlineSavingsMsg'
Learn More
4-(Trifluoroacetyl)toluene, 97%, Thermo Scientific Chemicals Product Code.: 11388795

4-(Trifluoroacetyl)toluene, 97%, Thermo Scientific Chemicals

Product Code. 11388795
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11388795

Brand: Thermo Scientific Alfa Aesar L12424.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 394-59-2
Molecular Formula C9H7F3O
Molecular Weight (g/mol) 188.149
MDL Number MFCD00052338
InChI Key DYILUJUELMWXAL-UHFFFAOYSA-N
Synonym 4-trifluoroacetyl toluene, 2,2,2-trifluoro-1-p-tolyl ethanone, 4'-methyl-2,2,2-trifluoroacetophenone, 2,2,2-trifluoro-1-p-tolylethanone, 2,2,2-trifluoro-1-4-methylphenyl ethanone, 2,2,2-trifluoro-4'-methylacetophenone, 2,2,2-trifluoro-1-4-methylphenyl ethan-1-one, 2,2,2-trifluoro-1-p-tolyl-ethanone, 4-trifluoroacetyltoluene, acmc-209j5mShow MoreShow Less
PubChem CID 136237
IUPAC Name 2,2,2-trifluoro-1-(4-methylphenyl)ethanone
SMILES CC1=CC=C(C=C1)C(=O)C(F)(F)F

Specifications

Boiling Point 70°C to 72°C (15 mmHg)
Refractive Index 1.47
Quantity 5 g
Beilstein 1870406
Formula Weight 188.15
Percent Purity 97%
Chemical Name or Material 4-(Trifluoroacetyl)toluene

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10202193 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberL12424.06,L12424.03
Lot Number10202193 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL12424.06,L12424.03
Lot Number10196293 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL12424.06,L12424.03
Lot Number10167940 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL12424.06,L12424.03
Lot Number10181773 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL12424.06,L12424.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-(Trifluoroacetyl)toluene, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.