Description
The 5X Kinase Buffer A is a concentrated general kinase assay buffer that can be used with a variety of assay technologies such as, LanthaScreen™, Adapta™, and Z’-LYTE™ assays.Ready to use and validated with LanthaScreen™, Adapta™, and Z’LYTE™ assays
Easily Screen for Kinase Targets using an Invitrogen™ Platform
For more information on the kinase assays that can be used with our 5X Kinase Buffer A see the following links:
LanthaScreen™ Eu Binding Assay
LanthaScreen™ Activity Assay
Adapta™ Universal Kinase Assay
Z´-LYTE™ Kinase Assay Kits
Save Freezer Space
The 5X Kinase Buffer can be stored at room temperature, saving you time and precious freezer space.
For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
Specifications
5X | |
5X Kinase Buffer A may be shipped frozen or at room temperature. It should be stored at room temperature (20-30°C). | |
Approved for shipment at Room Temperature or on Wet or Dry Ice | |
4 mL | |
Liquid |
Pharma & Biopharma; Protein Assays and Analysis | |
Biochemical binding/kinetic, Biochemical terbium phospho-specific activity, Biochemical europium phospho-specific activity | |
ABL1 G250E, CSF1R, CSK, EPHA5, KIT, PDGFRA T674I, PDGFRA V561D, PTK6, RET Y791F, SYK, TYRO3, ZAP70, MAP4K5, MST4, PRKCQ, PRKX, ROCK2, IKBKE, PAK4, EIF2AK1, MAP3K2, LRRK2 D1994A, GRK7, GSK3A, GSK3B, SNF1LK2, ACVR2A, BMPR1A, MAP2K1 S218D S222D, RAF1 (cRAF) Y340D Y341D, FGFR1, FGFR2, PRKCD, MAPK13, MAPK8, MAP3K10, MAP3K9, JAK1, JAK2 JH1 JH2, AMPK (A2/B1/G1), CAMK1, CAMK2B, CHEK1, MAPKAPK2, PASK, PIM2, PRKCN, PRKD2, PRKG2, RPS6KA5, MELK, STK33, BLK, EGFR, EPHB2, FGFR4, IGF1R, LYNA, MATK, MERTK, NTRK3, RET V804L, ROS1, TEK, TTK, NEK2, PRKCG, SGK, CHUK, IKBKB, TAOK3, JAK2 JH1 JH2 V617F, TYK2, DNAPK, IRAK1, ACVR1B, MAPKAPK5, MYLK2, SRPK2, STK4, WNK2, FLT1, MUSK, SRCN1, TEC, CAMK2D, MAP3K7/MAP3K7IP1, LRRK2 R1441C, CSNK1G1, DAPK2, DYRK1A, DYRK3, HIPK3, MAP2K1, MAP2K6, PIK3CG, RPS6KA6, DDR1, RIPK2, ABL1 E255K, BMX, EPHA4, FYN, PTK2, PTK2B, RET, MAP4K2, MINK1, SGK2, GRK6, MAPK9, JAK3, LRRK2, AMPK (A1/B1/G1), AURKA, CAMK1D, CAMK4, CHEK2, MAPKAPK3, PIM1, PKN1, PRKD1, PRKG1, RPS6KA4, STK22D, CSNK1G2, NUAK1, ABL1 T315I, EPHA3, EPHB4, FLT3, FLT4, INSR, LCK, YES1, EGFR L858R, ERBB4, KDR, MET, TNK2, PRKCA, PRKCI, CAMK2A, PAK1, RPS6KA3, MAPK14, EIF2AK3, MAP3K3, WNK3, ABL1, ABL2, AXL, FGFR3, FGFR3 K650E, FGR, HCK, KIT V654A, AURKB, TAOK2, MAP3K11, CAMKK1, CDC42BPA, CDK1, CDK5, CDK8, CDK9/Cyclin K, MAP2K3, TGFBR1, NLK, MAPK10, CSNK1G3, DMPK, DYRK1B, ACVR1, TGFBR2, EPHA1, EPHA8, EPHB3, INSRR, LYNB, MST1R, NTRK1, PDGFRA D842V, SRMS, MAP4K4, AKT2, LRRK2 G2019S, CAMKK2, CDC42BPB, CDK2, CDK7, CDK9/Cyclin T1, CSNK1E, EGFR L861Q, EPHA2, FER, LTK, PDGFRA, PDGFRB, MARK4, RPS6KB1, EGFR T790M, EPHA7, EPHB1, MET M1250T, DYRK4, PHKG1, PRKCZ, ROCK1, RPS6KA1, TBK1, MAPK1, MAPK11, BRAF V599E, JAK2, MARK1, PHKG2, MLCK, PAK6, SLK, LIMK1, MKNK2, MYLK, FRK, ITK, PRKCB1, STK24, AKT3, BRAF, LRRK2 I2020T, BRSK1, CLK2, PDK1, MAP2K2, PIK3CA/PIK3R1, PIK3CD/PIK3R1, ACVR2B, MAP3K14, MAP2K6 S207E T211E, FLT3 D835Y, SRC, GRK5, GRK4, STK3, ABL1 Y253F, BTK, DDR2, FES, KIT T670I, TXK, PRKCB2, RPS6KA2, SGKL, STK25, AKT1, PAK7, PRKACA, MAPK12, MAPK3, MAP3K8, CLK1, CLK4, MARK2, STK17A, ERBB2, NTRK2, PRKCE, PRKCH, EIF2AK2, MAP3K5, IRAK4, EGFR T790M L858R, LIMK2, STK16 | |
Kinase Buffer A |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 12
SelectScreen Kinase Profiling Service Adapta Protocol and Assay Conditions
File size: 337.2 kB
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
3028855C | Certificate of Analysis | 21/02/2025 | PV3189 |
2545572H | Certificate of Analysis | 21/02/2025 | PV3189 |
3028855B | Certificate of Analysis | 21/02/2025 | PV3189 |
3028855F | Certificate of Analysis | 21/02/2025 | PV3189 |
2545572D | Certificate of Analysis | 23/05/2023 | PV3189 |
Product Content Correction
Product Title
5X Kinase Buffer A > 4mL