missing translation for 'onlineSavingsMsg'
Learn More

6-Bromoimidazo[1,2-a]pyridine, 98%, Thermo Scientific Chemicals

36.38 EUR - 121.61 EUR

Chemical Identifiers

CAS 6188-23-4
Molecular Formula C7H5BrN2
Molecular Weight (g/mol) 197.035
MDL Number MFCD06496200
InChI Key FXPMFQUOGYGTAM-UHFFFAOYSA-N
Synonym 6-bromoimidazo 1,2-a pyridine, 6-bromoh-imidazo 1,2-a pyridine, 6-bromo-imidazo 1,2-a pyridine, imidazo 1,2-a pyridine, 6-bromo, buttpark 154\50-43, pubchem14736, acmc-209mx4, 6-bromo-4-hydroimidazo 1,2-a pyridineShow MoreShow Less
PubChem CID 2795547
IUPAC Name 6-bromoimidazo[1,2-a]pyridine
SMILES C1=CC2=NC=CN2C=C1Br
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15457397
SDS View Documents
Thermo Scientific Alfa Aesar
H34453.03
1 g
36.38 EUR
1g
Please to purchase this item. Need a web account? Register with us today!
15467397
SDS View Documents
Thermo Scientific Alfa Aesar
H34453.06
5 g
121.61 EUR
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is used in organic syntheses and as pharmaceutical intermediates. The best results were obtained with lactams that could be introduced on the 6-bromoimidazo[1,2-a]pyridine. 1,2-dihydro-5-imidazo [1,2-a]pyridin-6-yl-6-methyl-2-oxo-3-pyridinecarbonitrile hydrochloride monohydrate 6 was synthesized in 5 steps from 6-bromoimidazo[1,2-a] pyridine using [2- 14 C] cyanoacetamide as the source of the radiolabel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in organic syntheses and as pharmaceutical intermediates. The best results were obtained with lactams that could be introduced on the 6-bromoimidazo[1,2-a]pyridine. 1,2-dihydro-5-imidazo [1,2-a]pyridin-6-yl-6-methyl-2-oxo-3-pyridinecarbonitrile hydrochloride monohydrate 6 was synthesized in 5 steps from 6-bromoimidazo[1,2-a] pyridine using [2- 14 C] cyanoacetamide as the source of the radiolabel.

Solubility
slightly soluble in water.

Notes
Incompatible with strong oxidizing agents. Store at room temperature.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
6-Bromoimidazo[1,2-a]pyridine, 98%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.