Learn More
6-Hydroxy-3-iodo-2-methylpyridine, 95%, Thermo Scientific™ Product Code.: 11397169
6-Hydroxy-3-iodo-2-methylpyridine, 95%, Thermo Scientific™
Product Code. 11397169
1 g
Click to view available options
Quantity:
250 mg
1 g
Product Code. 11397169
Brand: Thermo Scientific Alfa Aesar H27381.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 927870-76-6
Molecular Formula C6H6INO
Molecular Weight (g/mol) 235.024
MDL Number MFCD09037468
InChI Key UFZSHCNPDKACCS-UHFFFAOYSA-N
Synonym 5-iodo-6-methylpyridin-2-ol, 5-iodo-6-methylpyridin-2 1h-one, 2-hydroxy-5-iodo-6-methylpyridine, 5-iodo-6-methyl-pyridin-2-ol, 3-iodo-6-hydroxy-2-methylpyridine, 5-iodo-6-methyl-1,2-dihydropyridin-2-one
PubChem CID 42553029
IUPAC Name 5-iodo-6-methyl-1H-pyridin-2-one
SMILES CC1=C(C=CC(=O)N1)I
Specifications
Melting Point ∽152°C (decomposition)
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 235.02
Percent Purity 95%
Chemical Name or Material 6-Hydroxy-3-iodo-2-methylpyridine
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10172080 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot Number10147942 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot NumberJ25Y002 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction
Product Title
6-Hydroxy-3-iodo-2-methylpyridine, 95%, Thermo Scientific™ >