missing translation for 'onlineSavingsMsg'
Learn More
6-Phenyl-2-hexyne, 99%, Thermo Scientific Chemicals Product Code.: 11400481

6-Phenyl-2-hexyne, 99%, Thermo Scientific Chemicals

Product Code. 11400481
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11400481

Brand: Thermo Scientific Alfa Aesar B20019.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34298-75-4
Molecular Formula C12H14
Molecular Weight (g/mol) 158.244
MDL Number MFCD00026982
InChI Key BYQGPSNMZZGRQP-UHFFFAOYSA-N
Synonym 6-phenyl-2-hexyne, hex-4-yn-1-ylbenzene, 1-phenyl-4-hexyne, acmc-20alxr, methyl 3-phenylpropyl acetylene
PubChem CID 3251198
IUPAC Name hex-4-ynylbenzene
SMILES CC#CCCCC1=CC=CC=C1

Specifications

Quantity 5 g
Formula Weight 158.25
Percent Purity 99%
Chemical Name or Material 6-Phenyl-2-hexyne

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10250368 Certificate TypeCertificate of Analysis Date08/04/2024 Catalog NumberB20019.03,B20019.06
Lot Number10244485 Certificate TypeCertificate of Analysis Date08/11/2022 Catalog NumberB20019.03,B20019.06
Lot Number10232927 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB20019.03,B20019.06
Lot Number10166842 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB20019.03,B20019.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
6-Phenyl-2-hexyne, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.