missing translation for 'onlineSavingsMsg'
Learn More
SHOWA™ 7585 Single-Use Nitrile Gloves, Powder-Free Product Code.: 16203172

SHOWA™ 7585 Single-Use Nitrile Gloves, Powder-Free

Product Code. 16203172
Small
Click to view available options
Size:
Small
Large
X-Large
2X-Large
This item is not returnable. View return policy

Product Code. 16203172

Brand: SHOWA™ 758507S

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Heavy duty, extra strong, highly penetration safe, long cuff single use chemical protective glove!

  • Thicker glove provides more resistance to chemicals 
  • Longer cuff for more protection
  • Avoids latex allergies risks type I and type IV
  • Second skin feel, softer texture and easy donning
  • Low-modulus formulation to improve fit and reduce hand fatigue
  • Textured finish on fingertips to enhance grip
  • Without plasticisers, powder-free and silicone-free

Specifications

Certifications/Compliance (Europe) EU Regulation 425/2016, EN 420, EN 455, EN 374-1:2016, EN 374-5:2016, EU Regulation 1935/2004 & 10/2011
AQL 0.65
Color Cobalt Blue
Disposable Single-use
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size Small
Sterility Non-sterile
Thickness (Metric) 0.20 mm
Finger Style Textured
Model 7585S
Packaging Type dispenser box
Port Size (Metric) 30 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
SHOWA™ 7585 Single-Use Nitrile Gloves, Powder-Free >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.