Learn More
Integra Biosciences™ 8-Channel Aspiration Adapter
Convert any vacuum source into a user-friendly aspiration system. Integra Biosciences™ 8-Channel Aspiration Adapters fit on the Vacuboy™ hand operator so that your vacuum source is converted into a flexible system for aspirating liquids from tubes, dishes, bottles, T-Flasks, 24-well and 96-well plates.
Brand:  Integra Biosciences™ 155520
Product Code. 10043471
  • 89.40 EUR / Each
Estimated Shipment: 16-05-2025
to see stock.
Explore more special offers
Description
VACUBOY hand operator set comprises of: VACUBOY hand operator, stand, rubber adapter for Pasteur pipettes, 1 channel stainless steel aspiration adapter 40mm, 8 channel stainless steel aspiration adapter 40mm, 1 channel plastic aspiration adapter for disposable tips, 1 channel plastic aspiration adapter for disposable tips with ejector, 8 channel plastic aspiration adapter for disposable tips with ejector. Can be used with VACUSAFE comfort vacuum system. Full range of accessories available. Enables liquid residues to be disposed of reliably, precisely and conveniently:
  • With a precisely dosed volume
  • Without loss of drops
  • Without risk of contamination
  • Without causing tiredness
  • Energy saving, the manual operator - the heart of the system - can be operated as a single or 8 channel aspirator
  • All parts in contact with fluids are autoclavable
Specifications
Specifications
8-channel Adapter
with ejector
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Integra Biosciences™ 8-Channel Aspiration Adapter > Product Type: 8-Channel Aspiration Adapter