Learn More
Tocris Bioscience™ AC 1903
Selective TRPC5 blocker
Brand: Tocris Bioscience™ 6766/5
This item is not returnable.
View return policy
Description
AC 1903 is a selective TRPC5 blocker (IC50 = 13.6 μM). Also displays weak inhibition at TRPC4 (IC50 >100 μM) but not TRPC6. Exhibits no effect against a standard panel of profiling kinases. Blocks TRPC5 channel activity in glomeruli of proteinuric rats. Suppresses severe proteinuria and prevents podocyte loss in a transgenic rat model of focal segmental glomerulosclerosis (FSGS).Specifications
N-(2-Furanylmethyl)-1-(phenylmethyl)-1H-benzimidazol-2-amine | |
5 mg | |
303.36 |
831234-13-0 | |
C19H17N3O | |
98% |