Learn More
BINDER™ Access Port
Access port with silicone plug for use with KB65, KB130, KB260, KB470, KB720, and KB 1060.
Brand:  BINDER™ 8012-2470
 View more versions of this product
Product Code. 30238224
  • 943.00 EUR / Each
Estimated Shipment: 30-04-2025
Description
  • Round access port with silicone plug
  • Choose between 30, 50, or 100 mm plug
  • Available on left, right, or top of the chamber
  • For use with KB65, KB130, KB260, KB470, KB720, and KB 1060
Specifications
Specifications
KB 65, KB 130, KB 260, KB 470, KB 720, KB 1060
Access Port
100 mm
Silicone
1 on Right Side
Product Title
BINDER™ Access Port > Access Port, 100 mm, Right