Learn More
Thermo Scientific™ Accessories for MaxQ™ HP Orbital Shakers
Get maximum flexibility, regardless of your application, by using these accessories for Thermo Scientific™ MaxQ™ High Performance orbital shakers.
Brand:  Thermo Scientific™ 452000
4294.40 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
 View more versions of this product
Product Code. 10474432
  • 4864.20 EUR / Each
Estimated Shipment: 19-06-2025
Description
Get maximum flexibility, regardless of your application, by using Thermo Scientific™ Accessories for MaxQ™ High Performance Orbital Shakers.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Shaker Kit with Mount and Stand
Dual-stack benchtop shaker kit; Includes kneewell side mount and orbital shaker stand
MaxQ™ HP 420 Shaker
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2112702 Certificate TypeCertificate of Analysis Date13/08/2019 Catalog Number452000
Lot Number1789023 Certificate TypeCertificate of Analysis Date23/01/2018 Catalog Number452000
Lot Number1883788 Certificate TypeCertificate of Analysis Date04/01/2018 Catalog Number452000
Lot Number1629744 Certificate TypeCertificate of Analysis Date19/07/2017 Catalog Number452000
Lot Number1228511 Certificate TypeCertificate of Analysis Date13/04/2017 Catalog Number452000
Product Title
Thermo Scientific™ Accessories for MaxQ™ HP Orbital Shakers > Dual-stack benchtop shaker kit; Includes kneewell side mount and orbital shaker stand; For model 420