Learn More
Cytiva Accessories for Multiphor™ II Electrophoresis System
Brand: Cytiva 1850-912
This item is not returnable.
View return policy
Description
Specifications
Standard flatbed unit | |
Multiphor II Electrophoresis System | |
EPH Electrode Wick (104 x 253 mm) |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Product Content Correction
Product Title
Cytiva Accessories for Multiphor™ II Electrophoresis System > EPH Electrode Wicks; 104 x 253mm
Spot an opportunity for improvement?Share a Content Correction