missing translation for 'onlineSavingsMsg'
Learn More

Acenaphthenequinone, 95%, Thermo Scientific Chemicals

Product Code. 11405467
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Acenaphthenequinone is used in the preparation of acetylcholinesterase inhibitory agents. It is also used in the preparation of turn-on sensors for cysteine/homocysteine and its application in bioimaging. Further, it is used as an intermediate in manufacturing of dyes and pharmaceuticals. In addition to this, it is used in chemical research as a drug and therapeutic agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acenaphthenequinone is used in the preparation of acetylcholinesterase inhibitory agents. It is also used in the preparation of turn-on sensors for cysteine/homocysteine and its application in bioimaging. Further, it is used as an intermediate in manufacturing of dyes and pharmaceuticals. In addition to this, it is used in chemical research as a drug and therapeutic agent.

Solubility
Soluble in chloroform and alcohol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 82-86-0
Molecular Formula C12H6O2
Molecular Weight (g/mol) 182.178
MDL Number MFCD00003805
InChI Key AFPRJLBZLPBTPZ-UHFFFAOYSA-N
Synonym acenaphthenequinone, acenaphthoquinone, 1,2-acenaphthylenedione, acenaphthenedione, 1,2-acenaphthenequinone, 1,2-acenaphthenedione, acenaphthaquinone, acenaphthylene-1,2-quinone, acenaphthylenequinone, acenaphthodione
PubChem CID 6724
ChEBI CHEBI:15342
IUPAC Name acenaphthylene-1,2-dione
SMILES C1=CC2=C3C(=C1)C(=O)C(=O)C3=CC=C2

Specifications

Melting Point 253°C to 262°C
Quantity 25 g
Beilstein 879172
Solubility Information Soluble in chloroform and alcohol. Insoluble in water.
Formula Weight 182.18
Percent Purity 95%
Chemical Name or Material Acenaphthenequinone
compliance-icons
Product Identifier
  • Acenaphthenequinone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 1,2-Acenaphthylenedione

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acenaphthenequinone, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.