missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile, puriss. p.a., ACS Reagent, Reag. Ph. Eur., ≥99.5% (GC), Honeywell Riedel-de Haën™
Click to view available options
Quantity:
4 x 2.5 L
6 x 1 L
155 kg
Packaging:
Glass Bottle
Metal Drum
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications

Specifications
Specifications
Melting Point | -48°C |
Acidity | Free acid (as CH3COOH): ≤0.001% |
Alkalinity | Free alkali (as NH3): ≤0.001% |
Density | 0.786g/mL at 25°C |
Boiling Point | 82°C |
Quantity | 4 x 2.5 L |
Explosion Limit | 0.16 |
Flash Point | 2°C (35.6°F) |
Ignition Point | 973°F |
Packaging | Glass Bottle |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Acetonitrile, puriss. p.a., ACS Reagent, Reag. Ph. Eur., ≥99.5% (GC), Honeywell Riedel-de Haën™ >
Spot an opportunity for improvement?Share a Content Correction