Learn More
Cytiva (formerly Pall Lab) Acrodisc™ Syringe Filters with Versapor™ Membrane, Sterile
Syringe filter for use in cell culture and other sensitive biological research.
Brand:  Cytiva (formerly Pall Lab) 4188
Additional Details : Weight : 0.32300kg
 View more versions of this product
Product Code. 17144271
  • 216.00 EUR / Pack of 50
Estimated Shipment: 24-01-2025
Description
  • Meets aqueous sample filtration needs
  • Designed for laboratory applications only
  • This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Specifications
Acrylic Copolymer (Hydrophilic)
Sterile
50/Pk.
0.8 μm
2.8 cm2
25 mm