Learn More
Cytiva (formerly Pall Lab) Acrodisc™ Syringe Filters with Versapor™ Membrane, Sterile
Syringe filter for use in cell culture and other sensitive biological research.
Brand: Cytiva (formerly Pall Lab) 4199
Additional Details : Weight : 0.30400kg
Description
- Meets aqueous sample filtration needs
- Designed for laboratory applications only
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Acrylic Copolymer (Hydrophilic) | |
Sterile | |
50/Pk. |
5 μm | |
2.8 cm2 | |
25 mm |