Learn More
Cytiva AcroPak™ 1500 Capsules with Supor™ Membrane
AcroPak Capsules for buffers, tissue culture media, and other biological fluids.
Brand: Cytiva 12686
269.80 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.
This item is not returnable.
View return policy
Description
- Newer technology recommended for simplified scale-up
- High filtration area and compact size are ideal for upscale trials
- Designed for research applications only
- Gamma-irradiated
Specifications
Hydrophilic PES | |
Sterile | |
1/2 in. Hose Barb |
0.2/0.2 μm | |
1500 cm2 | |
1/Ea. |
Product Content Correction
Product Title
Cytiva AcroPak™ 1500 Capsules with Supor™ Membrane > 0.2 μm/0.2 μm
Spot an opportunity for improvement?Share a Content Correction