missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAM19 Ectodomain Rat anti-Mouse, Clone: 164646, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB1229-SP

This item is not returnable.
View return policy
Description
ADAM19 Monoclonal specifically detects ADAM19 in Mouse samples. It is validated for Western Blot.Specifications
ADAM19 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
O35674 | |
ADAM19 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse ADAM19 Ectodomain Asp205-Gly705 (predicted) Accession # O35674 | |
25 μg | |
Primary | |
Detects mouse ADAM19 Ectodomain in direct ELISAs and Western blots. It also recognizes a C-terminal fragment (amino acids 588-705) of recombinant mouse (rm) ADAM19 in Western blots. In Western blots, no cross-reactivity with rmADAM10 or rmADAM15 is observed. | |
Mouse | |
Purified |
Western Blot | |
164646 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
a disintegrin and metalloproteinase domain 19 (meltrin beta), ADAM 19, ADAM metallopeptidase domain 19, disintegrin and metalloproteinase domain-containing protein 19, EC 3.4.24, EC 3.4.24.-, MADDAM, Meltrin beta, Meltrin-beta, Metalloprotease and disintegrin dendritic antigen marker, MLTNBmeltrin beta | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
8728 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
ADAM19 Ectodomain Rat anti-Mouse, Clone: 164646, R&D Systems™
Spot an opportunity for improvement?Share a Content Correction