Learn More
SCHOTT AG Lighting and Imaging™ Adapter for Transmitted Light Base (TLS)
This Adapter fits the transmitted light stage to a microscope stand with base plate.
Brand:  SCHOTT AG Lighting and Imaging™ 157630
 View more versions of this product
Product Code. 15743022
  • 73.55 EUR / 1 unit
Estimated Shipment: 28-02-2025
to see stock.
Explore more special offers
Description
  • Adapter for TLS 122150 Transmitted Light Base.
  • Models available to fit microscope stands with base plates that have diameters of 90 mm, 100 mm, 120 mm and 180 mm.
Specifications
Specifications
Adapter
90 mm
TLS 122150 Transmitted Light Base
3.5 in.
Stereo Microscopy Illumination