Learn More
Agar (80-100 Mesh), Fisher BioReagents
103.00 EUR - 185.00 EUR
Specifications
Form Solid
Product Type Agar
Packaging Type Poly Bottle
View More Specs
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10747024
View Documents
Fisher BioReagents
BP2641-100
100 g
103.00 EUR
100g
In Stock
to see stock.
Add to basket
11865465
View Documents
Fisher BioReagents
BP2641-500
500 g
185.00 EUR
500g
In Stock
to see stock.
Add to basket
10346693
View Documents Promotion Details
Fisher BioReagents
BP2641-1
1 kg
169.00 EUR
1kg
In Stock
Specifications
Solid
Agar
Poly Bottle